Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGMB
Entrez Gene ID
285704
Gene Name
repulsive guidance molecule family member b
Gene Aliases
DRAGON
Chromosome Location
Chr.5: 98768386 - 98796494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012761.2 NP_001012779.2 4 - 5 1168 68
XM_005271966.3 XP_005272023.1 4 - 5 1024 68
XM_011543345.2 XP_011541647.1 4 - 5 929 68
XM_011543346.2 XP_011541648.1 2 - 3 1306 68
XM_017009390.1 XP_016864879.1 4 - 5 1416 68
XM_017009391.1 XP_016864880.1 4 - 5 1314 68
GenBank mRNA AK125284.1 - 1 - 2 67 68

View Full Product Details