The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000131.4 | NP_000122.1 | 7 - 8 | 736 | 68 |
NM_001267554.1 | NP_001254483.1 | 4 - 5 | 484 | 68 | |
NM_019616.3 | NP_062562.1 | 6 - 7 | 670 | 68 | |
NR_051961.1 | 5 - 6 | 703 | 68 | ||
XM_006719963.3 | XP_006720026.2 | 5 - 6 | 544 | 68 | |
XM_011537474.2 | XP_011535776.2 | 7 - 8 | 793 | 68 | |
XM_011537475.2 | XP_011535777.2 | 5 - 6 | 607 | 68 | |
XM_011537476.2 | XP_011535778.1 | 4 - 5 | 1151 | 68 | |
GenBank mRNA | AK298404.1 | - | 4 - 5 | 481 | 68 |
AK311565.1 | - | 6 - 7 | 585 | 68 | |
AK311682.1 | - | 5 - 6 | 652 | 68 | |
BC130468.1 | - | 6 - 7 | 660 | 68 | |
EF421855.1 | - | 1 - 2 | 105 | 68 | |
EU557239.1 | - | 7 - 8 | 686 | 68 | |
M13232.1 | - | 7 - 8 | 717 | 68 |