Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNRG
Entrez Gene ID
11276
Gene Name
synergin, gamma
Gene Aliases
AP1GBP1, SYNG
Chromosome Location
Chr.17: 37514797 - 37613637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163544.2 NP_001157016.1 4 - 5 453 98
NM_001163545.2 NP_001157017.1 4 - 5 450 98
NM_001163546.2 NP_001157018.1 4 - 5 453 98
NM_001163547.2 NP_001157019.1 4 - 5 453 98
NM_007247.5 NP_009178.3 4 - 5 453 98
NM_080550.4 NP_542117.3 4 - 5 453 98
NM_198882.2 NP_942583.1 4 - 5 453 98
XM_005256980.4 XP_005257037.1 4 - 5 511 98
XM_017024076.1 XP_016879565.1 4 - 5 511 98
XM_017024077.1 XP_016879566.1 4 - 5 430 98
XM_017024078.1 XP_016879567.1 4 - 5 511 98
XM_017024079.1 XP_016879568.1 4 - 5 511 98
XM_017024080.1 XP_016879569.1 4 - 5 4563 98
XM_017024081.1 XP_016879570.1 4 - 5 4560 98
XM_017024082.1 XP_016879571.1 4 - 5 511 98
XM_017024083.1 XP_016879572.1 4 - 5 511 98
XM_017024084.1 XP_016879573.1 4 - 5 511 98
XM_017024085.1 XP_016879574.1 4 - 5 511 98
XM_017024086.1 XP_016879575.1 4 - 5 511 98
XM_017024087.1 XP_016879576.1 4 - 5 511 98
XM_017024088.1 XP_016879577.1 4 - 5 511 98
XM_017024089.1 XP_016879578.1 4 - 5 511 98
XM_017024090.1 XP_016879579.1 4 - 5 511 98
XM_017024091.1 XP_016879580.1 4 - 5 430 98
XM_017024092.1 XP_016879581.1 4 - 5 511 98
XM_017024093.1 XP_016879582.1 4 - 5 511 98
XM_017024094.1 XP_016879583.1 4 - 5 430 98
XM_017024095.1 XP_016879584.1 4 - 5 511 98
XM_017024096.1 XP_016879585.1 4 - 5 511 98
XM_017024097.1 XP_016879586.1 4 - 5 511 98
XM_017024098.1 XP_016879587.1 4 - 5 511 98
XM_017024099.1 XP_016879588.1 4 - 5 430 98
XM_017024100.1 XP_016879589.1 4 - 5 511 98
XM_017024101.1 XP_016879590.1 4 - 5 430 98
XM_017024102.1 XP_016879591.1 4 - 5 430 98
XM_017024103.1 XP_016879592.1 4 - 5 511 98
XM_017024104.1 XP_016879593.1 4 - 5 511 98
XM_017024105.1 XP_016879594.1 4 - 5 511 98
XM_017024106.1 XP_016879595.1 4 - 5 511 98
GenBank mRNA AF169548.1 - 4 - 5 413 98
AK126988.1 - 3 - 4 878 98
AK302821.1 - 4 - 5 455 98
BC090930.1 - 4 - 5 404 98
BC117313.1 - 4 - 5 393 98
BC143476.1 - 4 - 5 396 98
BC143478.1 - 4 - 5 396 98
DB085203.1 - 4 - 5 456 98

View Full Product Details