The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001438.2 | NP_001001438.1 | 1 - 2 | 90 | 70 |
NM_001145436.1 | NP_001138908.1 | 1 - 2 | 90 | 70 | |
NM_002340.5 | NP_002331.3 | 1 - 2 | 90 | 70 | |
XM_006724004.3 | XP_006724067.1 | 1 - 2 | 86 | 70 | |
XM_011529564.2 | XP_011527866.1 | 1 - 2 | 86 | 70 | |
XM_011529565.1 | XP_011527867.1 | 1 - 2 | 86 | 70 | |
XM_011529566.2 | XP_011527868.1 | 1 - 2 | 40 | 70 | |
XM_011529567.2 | XP_011527869.1 | 1 - 2 | 86 | 70 | |
XM_017028346.1 | XP_016883835.1 | 1 - 2 | 86 | 70 | |
XM_017028347.1 | XP_016883836.1 | 1 - 2 | 40 | 70 | |
XM_017028348.1 | XP_016883837.1 | 1 - 2 | 86 | 70 | |
GenBank mRNA | AK296313.1 | - | 1 - 2 | 47 | 70 |
AK312489.1 | - | 1 - 2 | 40 | 70 | |
BC035638.1 | - | 1 - 2 | 53 | 70 | |
BX443698.2 | - | 1 - 2 | 88 | 70 | |
D63807.1 | - | 1 - 2 | 29 | 70 | |
S81221.1 | - | 1 - 2 | 29 | 70 |