The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001438.2 | NP_001001438.1 | 22 - 23 | 2315 | 75 |
GenBank mRNA | D63807.1 | - | 22 - 23 | 2254 | 75 |
S81221.1 | - | 22 - 23 | 2254 | 75 |