The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001438.2 | NP_001001438.1 | 7 - 8 | 864 | 87 |
NM_001145436.1 | NP_001138908.1 | 7 - 8 | 831 | 87 | |
NM_001145437.1 | NP_001138909.1 | 6 - 7 | 996 | 87 | |
NM_002340.5 | NP_002331.3 | 7 - 8 | 864 | 87 | |
XM_006724004.3 | XP_006724067.1 | 7 - 8 | 860 | 87 | |
XM_011529564.2 | XP_011527866.1 | 7 - 8 | 860 | 87 | |
XM_011529565.1 | XP_011527867.1 | 7 - 8 | 860 | 87 | |
XM_011529566.2 | XP_011527868.1 | 7 - 8 | 781 | 87 | |
XM_011529567.2 | XP_011527869.1 | 7 - 8 | 860 | 87 | |
XM_017028346.1 | XP_016883835.1 | 7 - 8 | 860 | 87 | |
XM_017028347.1 | XP_016883836.1 | 7 - 8 | 781 | 87 | |
XM_017028348.1 | XP_016883837.1 | 7 - 8 | 860 | 87 | |
GenBank mRNA | AK226141.1 | - | 6 - 7 | 909 | 87 |
AK296313.1 | - | 7 - 8 | 788 | 87 | |
AK312489.1 | - | 7 - 8 | 814 | 87 | |
BC035638.1 | - | 7 - 8 | 827 | 87 | |
D63807.1 | - | 7 - 8 | 803 | 87 | |
S81221.1 | - | 7 - 8 | 803 | 87 | |
X87809.1 | - | 6 - 7 | 666 | 87 |