The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006545.4 | NP_006536.3 | 6 - 7 | 1085 | 67 |
XM_005264808.4 | XP_005264865.1 | 6 - 7 | 697 | 67 | |
XM_011533288.2 | XP_011531590.1 | 5 - 6 | 767 | 67 | |
XM_017005555.1 | XP_016861044.1 | 6 - 7 | 800 | 67 | |
XM_017005556.1 | XP_016861045.1 | 5 - 6 | 781 | 67 | |
GenBank mRNA | AF040707.1 | - | 6 - 7 | 745 | 67 |
AF040708.1 | - | 5 - 6 | 565 | 67 | |
AK292196.1 | - | 6 - 7 | 744 | 67 | |
AK309658.1 | - | 5 - 6 | 949 | 67 | |
AL833426.1 | - | 4 - 5 | 1436 | 67 | |
BC021984.1 | - | 6 - 7 | 831 | 67 | |
BC056861.1 | - | 6 - 7 | 783 | 67 | |
CR542035.1 | - | 3 - 4 | 322 | 67 |