Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBF2
Entrez Gene ID
81846
Gene Name
SET binding factor 2
Gene Aliases
CMT4B2, DENND7B, MTMR13
Chromosome Location
Chr.11: 9778667 - 10294216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030962.3 NP_112224.1 31 - 32 4397 88
XM_005253154.4 XP_005253211.1 32 - 33 4502 88
XM_005253155.4 XP_005253212.1 31 - 32 4370 88
XM_011520394.2 XP_011518696.1 31 - 32 4385 88
XM_017018372.1 XP_016873861.1 31 - 32 5791 88
XM_017018373.1 XP_016873862.1 32 - 33 4421 88
XM_017018374.1 XP_016873863.1 30 - 31 4277 88
XM_017018375.1 XP_016873864.1 30 - 31 4265 88
GenBank mRNA AY234241.1 - 31 - 32 4363 88
BC101466.1 - 31 - 32 4356 88
BX647533.1 - 4 - 5 3974 88
CR749312.1 - 5 - 6 701 88

View Full Product Details