The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143885.1 | NP_001137357.1 | 12 - 13 | 1677 | 162 |
NM_001143886.1 | NP_001137358.1 | 11 - 12 | 1432 | 162 | |
NM_001244990.1 | NP_001231919.1 | 12 - 13 | 1677 | 162 | |
NM_002480.2 | NP_002471.1 | 11 - 12 | 1821 | 162 | |
XM_005268887.3 | XP_005268944.1 | 11 - 12 | 1820 | 162 | |
XM_011538374.1 | XP_011536676.1 | 12 - 13 | 1927 | 162 | |
XM_011538376.2 | XP_011536678.1 | 12 - 13 | 1927 | 162 | |
XM_011538377.1 | XP_011536679.1 | 11 - 12 | 1852 | 162 | |
XM_011538378.2 | XP_011536680.1 | 12 - 13 | 1927 | 162 | |
XM_011538380.1 | XP_011536682.1 | 12 - 13 | 1927 | 162 | |
XM_011538383.2 | XP_011536685.1 | 11 - 12 | 1810 | 162 | |
XM_017019324.1 | XP_016874813.1 | 10 - 11 | 1735 | 162 | |
XM_017019329.1 | XP_016874818.1 | 10 - 11 | 1735 | 162 | |
XM_017019330.1 | XP_016874819.1 | 12 - 13 | 1927 | 162 | |
GenBank mRNA | AF458589.1 | - | 12 - 13 | 1677 | 162 |
AK302692.1 | - | 11 - 12 | 1432 | 162 | |
AK314193.1 | - | 11 - 12 | 1561 | 162 | |
BC013941.1 | - | 2 - 3 | 187 | 162 | |
BC047898.1 | - | 11 - 12 | 1744 | 162 | |
BC092481.1 | - | 12 - 13 | 1677 | 162 | |
BC111752.1 | - | 11 - 12 | 1554 | 162 | |
D87930.1 | - | 11 - 12 | 1554 | 162 |