Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R12A
Entrez Gene ID
4659
Gene Name
protein phosphatase 1 regulatory subunit 12A
Gene Aliases
M130, MBS, MYPT1
Chromosome Location
Chr.12: 79773563 - 79935455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143885.1 NP_001137357.1 20 - 21 2765 107
NM_001143886.1 NP_001137358.1 19 - 20 2520 107
NM_001244990.1 NP_001231919.1 20 - 21 2765 107
NM_001244992.1 NP_001231921.1 18 - 19 2494 107
NM_002480.2 NP_002471.1 19 - 20 2909 107
XM_005268887.3 XP_005268944.1 19 - 20 2908 107
XM_011538374.1 XP_011536676.1 20 - 21 3015 107
XM_011538375.1 XP_011536677.1 20 - 21 2979 107
XM_011538376.2 XP_011536678.1 20 - 21 3015 107
XM_011538377.1 XP_011536679.1 19 - 20 2940 107
XM_011538378.2 XP_011536680.1 20 - 21 3015 107
XM_011538379.1 XP_011536681.1 19 - 20 2847 107
XM_011538380.1 XP_011536682.1 19 - 20 2838 107
XM_011538383.2 XP_011536685.1 18 - 19 2721 107
XM_011538384.1 XP_011536686.1 18 - 19 2670 107
XM_011538385.1 XP_011536687.1 17 - 18 2553 107
XM_017019323.1 XP_016874812.1 19 - 20 2862 107
XM_017019324.1 XP_016874813.1 18 - 19 2823 107
XM_017019325.1 XP_016874814.1 18 - 19 2787 107
XM_017019326.1 XP_016874815.1 18 - 19 2685 107
XM_017019327.1 XP_016874816.1 18 - 19 2730 107
XM_017019328.1 XP_016874817.1 17 - 18 2655 107
XM_017019329.1 XP_016874818.1 17 - 18 2646 107
GenBank mRNA AF458589.1 - 20 - 21 2765 107
AK302692.1 - 19 - 20 2520 107
AK314193.1 - 19 - 20 2649 107
AY380574.1 - 18 - 19 2474 107
BC111752.1 - 19 - 20 2642 107
D87930.1 - 19 - 20 2642 107

View Full Product Details