Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R12A
Entrez Gene ID
4659
Gene Name
protein phosphatase 1 regulatory subunit 12A
Gene Aliases
M130, MBS, MYPT1
Chromosome Location
Chr.12: 79773563 - 79935455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143885.1 NP_001137357.1 25 - 26 3122 85
NM_001143886.1 NP_001137358.1 24 - 25 2877 85
NM_001244990.1 NP_001231919.1 24 - 25 3017 85
NM_001244992.1 NP_001231921.1 23 - 24 2851 85
NM_002480.2 NP_002471.1 24 - 25 3266 85
XM_005268887.3 XP_005268944.1 25 - 26 3296 85
XM_011538374.1 XP_011536676.1 25 - 26 3372 85
XM_011538375.1 XP_011536677.1 25 - 26 3336 85
XM_011538376.2 XP_011536678.1 26 - 27 3403 85
XM_011538377.1 XP_011536679.1 24 - 25 3297 85
XM_011538379.1 XP_011536681.1 24 - 25 3204 85
XM_011538380.1 XP_011536682.1 24 - 25 3195 85
XM_011538383.2 XP_011536685.1 23 - 24 3078 85
XM_011538384.1 XP_011536686.1 23 - 24 3027 85
XM_011538385.1 XP_011536687.1 22 - 23 2910 85
XM_017019323.1 XP_016874812.1 24 - 25 3219 85
XM_017019324.1 XP_016874813.1 23 - 24 3180 85
XM_017019325.1 XP_016874814.1 23 - 24 3144 85
XM_017019326.1 XP_016874815.1 23 - 24 3042 85
XM_017019327.1 XP_016874816.1 24 - 25 3118 85
XM_017019328.1 XP_016874817.1 22 - 23 3012 85
XM_017019329.1 XP_016874818.1 22 - 23 3003 85
GenBank mRNA AF458589.1 - 24 - 25 3017 85
AK057712.1 - 5 - 6 2974 85
AK125671.1 - 1 - 2 2189 85
AK302692.1 - 24 - 25 2877 85
AK314193.1 - 24 - 25 3006 85
AY380574.1 - 23 - 24 2831 85
BC111752.1 - 24 - 25 2999 85
D87930.1 - 24 - 25 2999 85

View Full Product Details