Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCML2
Entrez Gene ID
10389
Gene Name
sex comb on midleg-like 2 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.X: 18239313 - 18355089 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006089.2 NP_006080.1 7 - 8 886 94
NR_033717.1 7 - 8 886 94
XM_006724458.1 XP_006724521.1 7 - 8 850 94
XM_017029219.1 XP_016884708.1 7 - 8 782 94
XM_017029220.1 XP_016884709.1 7 - 8 778 94
XM_017029221.1 XP_016884710.1 7 - 8 920 94
GenBank mRNA AK303064.1 - 6 - 7 752 94
AK315116.1 - 7 - 8 850 94
BC051913.1 - 7 - 8 828 94
BC064617.1 - 7 - 8 843 94
Y18004.1 - 7 - 8 820 94

View Full Product Details