Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIPA1L1
Entrez Gene ID
26037
Gene Name
signal induced proliferation associated 1 like 1
Gene Aliases
E6TP1
Chromosome Location
Chr.14: 71320418 - 71741229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284245.1 NP_001271174.1 19 - 20 5488 67
NM_015556.2 NP_056371.1 20 - 21 5551 67
XM_005267516.4 XP_005267573.1 20 - 21 6140 67
XM_011536630.1 XP_011534932.1 24 - 25 5902 67
XM_011536631.1 XP_011534933.1 24 - 25 6313 67
XM_011536632.1 XP_011534934.1 23 - 24 5822 67
XM_011536635.2 XP_011534937.1 23 - 24 5969 67
XM_011536638.2 XP_011534940.1 19 - 20 4625 67
XM_017021159.1 XP_016876648.1 26 - 27 6059 67
XM_017021160.1 XP_016876649.1 24 - 25 5981 67
XM_017021161.1 XP_016876650.1 25 - 26 5934 67
XM_017021162.1 XP_016876651.1 27 - 28 6116 67
XM_017021163.1 XP_016876652.1 22 - 23 5783 67
XM_017021164.1 XP_016876653.1 22 - 23 5790 67
XM_017021165.1 XP_016876654.1 24 - 25 5859 67
XM_017021166.1 XP_016876655.1 25 - 26 5992 67
XM_017021167.1 XP_016876656.1 24 - 25 5867 67
XM_017021168.1 XP_016876657.1 24 - 25 5841 67
XM_017021169.1 XP_016876658.1 23 - 24 5994 67
XM_017021170.1 XP_016876659.1 23 - 24 6352 67
XM_017021171.1 XP_016876660.1 24 - 25 5925 67
XM_017021172.1 XP_016876661.1 23 - 24 5800 67
XM_017021173.1 XP_016876662.1 22 - 23 5927 67
XM_017021174.1 XP_016876663.1 22 - 23 6167 67
XM_017021175.1 XP_016876664.1 22 - 23 6285 67
XM_017021176.1 XP_016876665.1 27 - 28 6112 67
XM_017021177.1 XP_016876666.1 27 - 28 6101 67
XM_017021178.1 XP_016876667.1 25 - 26 5939 67
XM_017021179.1 XP_016876668.1 24 - 25 5842 67
XM_017021180.1 XP_016876669.1 28 - 29 6154 67
XM_017021186.1 XP_016876675.1 25 - 26 5996 67
XM_017021187.1 XP_016876676.1 23 - 24 5862 67
XM_017021188.1 XP_016876677.1 22 - 23 5737 67
XM_017021189.1 XP_016876678.1 24 - 25 5846 67
XM_017021194.1 XP_016876683.1 25 - 26 5912 67
XM_017021195.1 XP_016876684.1 24 - 25 5849 67
XM_017021197.1 XP_016876686.1 25 - 26 4259 67
XM_017021198.1 XP_016876687.1 22 - 23 4000 67
XM_017021199.1 XP_016876688.1 26 - 27 4301 67
GenBank mRNA AF090989.1 - 19 - 20 5475 67
AF090990.1 - 20 - 21 5538 67
CR936651.1 - 5 - 6 1284 67

View Full Product Details