Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SIPA1L1
- Entrez Gene ID
-
26037
- Gene Name
- signal induced proliferation associated 1 like 1
- Gene Aliases
-
E6TP1
- Chromosome Location
-
Chr.14: 71320418 - 71741229 on Build GRCh38
View Full Product Details