Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX4
Entrez Gene ID
8723
Gene Name
sorting nexin 4
Gene Aliases
ATG24B
Chromosome Location
Chr.3: 125446644 - 125520214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003794.3 NP_003785.1 7 - 8 774 107
NR_073435.1 6 - 7 652 107
GenBank mRNA AF065485.1 - 8 - 9 732 107
AF130078.1 - 2 - 3 141 107
AK001835.1 - 7 - 8 736 107
AK298972.1 - 6 - 7 619 107
BC018762.1 - 7 - 8 750 107

View Full Product Details