The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003794.3 | NP_003785.1 | 8 - 9 | 826 | 142 |
NR_073435.1 | 7 - 8 | 704 | 142 | ||
GenBank mRNA | AF065485.1 | - | 9 - 10 | 784 | 142 |
AF130078.1 | - | 3 - 4 | 193 | 142 | |
AK001835.1 | - | 8 - 9 | 788 | 142 | |
AK298972.1 | - | 7 - 8 | 671 | 142 | |
BC018762.1 | - | 8 - 9 | 802 | 142 |