The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018353.4 | NP_060823.3 | 10 - 11 | 2299 | 194 |
XM_005267833.4 | XP_005267890.1 | 11 - 12 | 2346 | 194 | |
XM_011536923.1 | XP_011535225.1 | 7 - 8 | 784 | 194 | |
XM_017021425.1 | XP_016876914.1 | 12 - 13 | 2327 | 194 | |
GenBank mRNA | AB067490.1 | - | 10 - 11 | 2300 | 194 |
AJ242977.1 | - | 3 - 4 | 276 | 194 | |
BC065544.1 | - | 10 - 11 | 2090 | 194 |