The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004581.5 | NP_004572.3 | 14 - 15 | 1982 | 164 |
NM_182836.2 | NP_878256.1 | 15 - 16 | 1665 | 164 | |
GenBank mRNA | AK291347.1 | - | 15 - 16 | 1549 | 164 |
AK292613.1 | - | 14 - 15 | 1714 | 164 | |
BC003093.1 | - | 15 - 16 | 1603 | 164 | |
CD370056.1 | - | 2 - 3 | 164 | ||
CR541797.1 | - | 13 - 14 | 1470 | 164 | |
Y08200.1 | - | 14 - 15 | 1744 | 164 |