The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014516.3 | NP_055331.1 | 16 - 17 | 2358 | 67 |
XM_005278279.1 | XP_005278336.1 | 16 - 17 | 2361 | 67 | |
XM_005278280.2 | XP_005278337.1 | 16 - 17 | 2395 | 67 | |
XM_005278281.1 | XP_005278338.1 | 16 - 17 | 2358 | 67 | |
XM_005278282.2 | XP_005278339.1 | 16 - 17 | 2361 | 67 | |
XM_011526992.1 | XP_011525294.1 | 16 - 17 | 2361 | 67 | |
GenBank mRNA | AB014591.1 | - | 15 - 16 | 3632 | 67 |
AK160386.1 | - | 15 - 16 | 2543 | 67 | |
BC016474.1 | - | 16 - 17 | 2311 | 67 |