The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002358.3 | NP_002349.1 | 4 - 5 | 568 | 102 |
GenBank mRNA | AA443733.1 | - | 2 - 3 | 297 | 102 |
AF394735.1 | - | 3 - 4 | 340 | 102 | |
AJ000186.1 | - | 4 - 5 | 510 | 102 | |
AK223433.1 | - | 4 - 5 | 529 | 102 | |
AK298228.1 | - | 3 - 4 | 422 | 102 | |
AK313827.1 | - | 4 - 5 | 530 | 102 | |
BC000356.2 | - | 4 - 5 | 518 | 102 | |
BC005945.1 | - | 4 - 5 | 511 | 102 | |
BC070283.1 | - | 4 - 5 | 480 | 102 |