Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
Chromosome Location
Chr.10: 59245933 - 59363186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001971.2 NP_001001971.1 6 - 7 721 77
NM_001143773.1 NP_001137245.1 7 - 8 892 77
NM_001166698.1 NP_001160170.1 7 - 8 806 77
NM_198215.3 NP_937858.2 6 - 7 721 77
XM_005269618.4 XP_005269675.2 6 - 7 721 77
XM_005269619.4 XP_005269676.2 6 - 7 721 77
XM_006717702.3 XP_006717765.1 8 - 9 722 77
XM_006717703.3 XP_006717766.1 8 - 9 904 77
XM_011539490.2 XP_011537792.1 7 - 8 777 77
XM_011539491.2 XP_011537793.1 8 - 9 823 77
XM_017015885.1 XP_016871374.1 7 - 8 635 77
XM_017015886.1 XP_016871375.1 7 - 8 892 77
XM_017015887.1 XP_016871376.1 7 - 8 642 77
XM_017015888.1 XP_016871377.1 8 - 9 904 77
XM_017015889.1 XP_016871378.1 8 - 9 722 77
XM_017015890.1 XP_016871379.1 8 - 9 961 77
XM_017015891.1 XP_016871380.1 7 - 8 892 77
XM_017015892.1 XP_016871381.1 7 - 8 892 77
XM_017015893.1 XP_016871382.1 9 - 10 993 77
GenBank mRNA AK289796.1 - 6 - 7 647 77
AK294805.1 - 6 - 7 636 77
AK299924.1 - 7 - 8 892 77
AK316542.1 - 7 - 8 802 77
BC036453.1 - 6 - 7 679 77
BC064431.1 - 6 - 7 721 77
U79304.1 - 4 - 5 329 77

View Full Product Details