The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135597.1 | NP_001129069.1 | 13 - 14 | 2363 | 146 |
NM_001254943.1 | NP_001241872.1 | 13 - 14 | 2363 | 146 | |
NM_018084.4 | NP_060554.3 | 13 - 14 | 2363 | 146 | |
XM_005264418.4 | XP_005264475.1 | 13 - 14 | 2066 | 146 | |
XM_005264421.2 | XP_005264478.1 | 13 - 14 | 2066 | 146 | |
XM_005264426.2 | XP_005264483.1 | 13 - 14 | 2066 | 146 | |
XM_011532965.2 | XP_011531267.1 | 13 - 14 | 2066 | 146 | |
XM_011532966.2 | XP_011531268.1 | 13 - 14 | 2066 | 146 | |
XM_011532967.2 | XP_011531269.1 | 13 - 14 | 2066 | 146 | |
XM_011532968.2 | XP_011531270.1 | 13 - 14 | 2066 | 146 | |
XM_017004476.1 | XP_016859965.1 | 13 - 14 | 2066 | 146 | |
XM_017004477.1 | XP_016859966.1 | 13 - 14 | 2066 | 146 | |
XM_017004478.1 | XP_016859967.1 | 13 - 14 | 2066 | 146 | |
XM_017004479.1 | XP_016859968.1 | 13 - 14 | 2066 | 146 | |
XM_017004480.1 | XP_016859969.1 | 11 - 12 | 1841 | 146 | |
XM_017004481.1 | XP_016859970.1 | 13 - 14 | 2066 | 146 | |
GenBank mRNA | AB125644.1 | - | 13 - 14 | 1521 | 146 |
AB201172.1 | - | 13 - 14 | 2055 | 146 | |
AF112218.1 | - | 13 - 14 | 2054 | 146 | |
AK307964.1 | - | 13 - 14 | 1761 | 146 | |
BC015539.1 | - | 3 - 4 | 354 | 146 | |
BC060037.1 | - | 13 - 14 | 1900 | 146 | |
BC078168.1 | - | 13 - 14 | 1513 | 146 | |
BC132736.1 | - | 13 - 14 | 1718 | 146 | |
BC144320.1 | - | 13 - 14 | 1718 | 146 | |
D64159.1 | - | 13 - 14 | 2054 | 146 |