The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135597.1 | NP_001129069.1 | 20 - 21 | 4440 | 100 |
NM_001254943.1 | NP_001241872.1 | 20 - 21 | 4440 | 100 | |
NM_018084.4 | NP_060554.3 | 20 - 21 | 4443 | 100 | |
XM_005264418.4 | XP_005264475.1 | 20 - 21 | 4146 | 100 | |
XM_005264421.2 | XP_005264478.1 | 20 - 21 | 4146 | 100 | |
XM_005264426.2 | XP_005264483.1 | 20 - 21 | 4146 | 100 | |
XM_011532965.2 | XP_011531267.1 | 20 - 21 | 4146 | 100 | |
XM_011532966.2 | XP_011531268.1 | 20 - 21 | 4146 | 100 | |
XM_011532967.2 | XP_011531269.1 | 20 - 21 | 4146 | 100 | |
XM_011532968.2 | XP_011531270.1 | 20 - 21 | 4146 | 100 | |
XM_017004476.1 | XP_016859965.1 | 20 - 21 | 4143 | 100 | |
XM_017004477.1 | XP_016859966.1 | 20 - 21 | 4146 | 100 | |
XM_017004478.1 | XP_016859967.1 | 20 - 21 | 4146 | 100 | |
XM_017004479.1 | XP_016859968.1 | 20 - 21 | 4146 | 100 | |
XM_017004480.1 | XP_016859969.1 | 18 - 19 | 3921 | 100 | |
XM_017004481.1 | XP_016859970.1 | 20 - 21 | 4146 | 100 | |
GenBank mRNA | AB033038.2 | - | 3 - 4 | 480 | 100 |
AB125644.1 | - | 20 - 21 | 3601 | 100 | |
AB201172.1 | - | 20 - 21 | 4132 | 100 | |
AK001254.1 | - | 3 - 4 | 542 | 100 | |
BC032683.1 | - | 4 - 5 | 747 | 100 | |
BC056885.1 | - | 5 - 6 | 877 | 100 | |
BC132736.1 | - | 20 - 21 | 3798 | 100 | |
BC144320.1 | - | 20 - 21 | 3795 | 100 | |
BX537985.1 | - | 6 - 7 | 1126 | 100 | |
BX538154.1 | - | 6 - 7 | 1049 | 100 | |
BX648138.1 | - | 6 - 7 | 1171 | 100 |