Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SP2
Entrez Gene ID
6668
Gene Name
Sp2 transcription factor
Gene Aliases
-
Chromosome Location
Chr.17: 47896032 - 47932271 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006722026.2 XP_006722089.1 2 - 3 228 107
XM_017024968.1 XP_016880457.1 3 - 4 274 107
GenBank mRNA D28588.1 - 1 - 2 323 107

View Full Product Details