The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033277.1 | NP_150593.1 | 4 - 5 | 388 | 99 |
GenBank mRNA | BC062217.1 | - | 4 - 5 | 408 | 99 |
BC069317.1 | - | 4 - 5 | 388 | 99 |