Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOG
Entrez Gene ID
4340
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
BTN6, BTNL11, MOGIG2, NRCLP7
Chromosome Location
Chr.6: 29656981 - 29672372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206809.3 NP_996532.2 6 - 7 941 88
NM_206814.5 NP_996537.3 5 - 6 593 88
GenBank mRNA AB209815.1 - 6 - 7 851 88
AK295213.1 - 7 - 8 762 88
AK308949.1 - 5 - 6 571 88
U64564.1 - 6 - 7 874 88
U64570.1 - 6 - 7 763 88

View Full Product Details