The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002161.5 | NP_002152.2 | 19 - 20 | 2169 | 125 |
NM_013417.3 | NP_038203.2 | 19 - 20 | 2339 | 125 | |
NR_073446.1 | 18 - 19 | 2012 | 125 | ||
GenBank mRNA | AB209234.1 | - | 19 - 20 | 2078 | 125 |
AK293014.1 | - | 19 - 20 | 2096 | 125 | |
BC065552.1 | - | 18 - 19 | 2011 | 125 | |
BX537417.1 | - | 19 - 20 | 2107 | 125 | |
BX538109.1 | - | 18 - 19 | 1952 | 125 | |
BX647071.1 | - | 19 - 20 | 2108 | 125 | |
BX647236.1 | - | 19 - 20 | 2101 | 125 | |
BX647430.1 | - | 19 - 20 | 2103 | 125 | |
BX648567.1 | - | 19 - 20 | 2109 | 125 | |
D28473.1 | - | 19 - 20 | 2257 | 125 | |
U04953.1 | - | 19 - 20 | 2271 | 125 |