The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037304.1 | NP_001032381.1 | 1 - 2 | 67 | 95 |
NM_001037305.1 | NP_001032382.1 | 1 - 2 | 67 | 95 | |
XM_005271527.3 | XP_005271584.1 | 1 - 2 | 102 | 95 | |
XM_005271528.3 | XP_005271585.1 | 1 - 2 | 102 | 95 | |
GenBank mRNA | BC093074.1 | - | 1 - 2 | 42 | 95 |
BQ640137.1 | - | 1 - 2 | 57 | 95 | |
BU934600.1 | - | 1 - 2 | 69 | 95 |