The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017890.4 | NP_060360.3 | 48 - 49 | 8976 | 83 |
NM_152564.4 | NP_689777.3 | 48 - 49 | 8901 | 83 | |
XM_005250800.3 | XP_005250857.1 | 48 - 49 | 9282 | 83 | |
XM_005250801.4 | XP_005250858.1 | 48 - 49 | 15152 | 83 | |
XM_011516848.2 | XP_011515150.1 | 48 - 49 | 8965 | 83 | |
XM_011516849.2 | XP_011515151.1 | 48 - 49 | 8890 | 83 | |
XM_011516850.2 | XP_011515152.1 | 46 - 47 | 8608 | 83 | |
XM_011516851.2 | XP_011515153.1 | 28 - 29 | 5868 | 83 | |
XM_011516852.2 | XP_011515154.1 | 28 - 29 | 5906 | 83 | |
XM_011516854.2 | XP_011515156.1 | 24 - 25 | 5064 | 83 | |
XM_017013109.1 | XP_016868598.1 | 47 - 48 | 8773 | 83 | |
XM_017013110.1 | XP_016868599.1 | 41 - 42 | 7766 | 83 | |
XM_017013111.1 | XP_016868600.1 | 28 - 29 | 6138 | 83 | |
XM_017013112.1 | XP_016868601.1 | 25 - 26 | 5280 | 83 | |
GenBank mRNA | AB011104.1 | - | 9 - 10 | 1710 | 83 |
AJ608772.1 | - | 48 - 49 | 8968 | 83 | |
AJ608773.1 | - | 48 - 49 | 8893 | 83 | |
AY223814.1 | - | 48 - 49 | 8976 | 83 | |
AY223815.1 | - | 48 - 49 | 8901 | 83 |