The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015725.2 | NP_056540.2 | 1 - 2 | 359 | 60 |
GenBank mRNA | AF229845.1 | - | 1 - 2 | 359 | 60 |
AK024022.1 | - | 1 - 2 | 125 | 60 |