The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204.6 | NP_001195.2 | 4 - 5 | 1676 | 108 |
XM_011511687.1 | XP_011509989.1 | 4 - 5 | 1693 | 108 | |
GenBank mRNA | AK292430.1 | - | 4 - 5 | 940 | 108 |
BC052985.2 | - | 4 - 5 | 1102 | 108 | |
D50516.1 | - | 4 - 5 | 936 | 108 | |
U20165.1 | - | 4 - 5 | 564 | 108 | |
Z48923.1 | - | 4 - 5 | 909 | 108 |