Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMPR2
Entrez Gene ID
659
Gene Name
bone morphogenetic protein receptor type 2
Gene Aliases
BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
Chromosome Location
Chr.2: 202376310 - 202567751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204.6 NP_001195.2 5 - 6 1771 109
XM_011511687.1 XP_011509989.1 5 - 6 1788 109
GenBank mRNA AK292430.1 - 5 - 6 1035 109
BC052985.2 - 5 - 6 1197 109
D50516.1 - 5 - 6 1031 109
U20165.1 - 5 - 6 659 109
Z48923.1 - 5 - 6 1004 109

View Full Product Details