The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204.6 | NP_001195.2 | 9 - 10 | 2427 | 132 |
XM_011511687.1 | XP_011509989.1 | 9 - 10 | 2444 | 132 | |
GenBank mRNA | AK292430.1 | - | 9 - 10 | 1691 | 132 |
BC052985.2 | - | 9 - 10 | 1853 | 132 | |
D50516.1 | - | 9 - 10 | 1687 | 132 | |
U20165.1 | - | 9 - 10 | 1315 | 132 | |
Z48923.1 | - | 9 - 10 | 1660 | 132 |