The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032861.3 | NP_116250.3 | 9 - 10 | 1000 | 96 |
XM_006715586.2 | XP_006715649.1 | 8 - 9 | 929 | 96 | |
XM_011536196.2 | XP_011534498.1 | 8 - 9 | 1023 | 96 | |
XM_011536198.2 | XP_011534500.1 | 7 - 8 | 925 | 96 | |
GenBank mRNA | AK027823.1 | - | 9 - 10 | 990 | 96 |
AK125878.1 | - | 10 - 11 | 2192 | 96 | |
BC001705.1 | - | 9 - 10 | 957 | 96 | |
BX648104.1 | - | 8 - 9 | 932 | 96 |