The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018465.3 | NP_060935.2 | 1 - 2 | 150 | 97 |
GenBank mRNA | AF225420.1 | - | 1 - 2 | 91 | 97 |
AK027594.1 | - | 1 - 2 | 89 | 97 | |
AK312736.1 | - | 1 - 2 | 79 | 97 | |
BC008212.1 | - | 1 - 2 | 60 | 97 | |
BM693747.1 | - | 1 - 2 | 150 | 97 |