Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPP2
Entrez Gene ID
7174
Gene Name
tripeptidyl peptidase 2
Gene Aliases
TPP-2, TPPII
Chromosome Location
Chr.13: 102596936 - 102679174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003291.2 NP_003282.2 17 - 18 2279 72
XM_005254070.1 XP_005254127.1 17 - 18 2278 72
XM_005254071.1 XP_005254128.1 17 - 18 2278 72
XM_005254072.1 XP_005254129.1 17 - 18 2278 72
GenBank mRNA AK304313.1 - 17 - 18 2187 72
BC039905.1 - 18 - 19 2222 72
M55169.1 - 17 - 18 2011 72
M73047.1 - 17 - 18 2199 72

View Full Product Details