The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003291.2 | NP_003282.2 | 27 - 28 | 3644 | 119 |
XM_005254070.1 | XP_005254127.1 | 29 - 30 | 3808 | 119 | |
XM_005254071.1 | XP_005254128.1 | 28 - 29 | 3769 | 119 | |
XM_005254072.1 | XP_005254129.1 | 28 - 29 | 3682 | 119 | |
GenBank mRNA | AK097678.1 | - | 6 - 7 | 1359 | 119 |
AK127549.1 | - | 4 - 5 | 3138 | 119 | |
BC039905.1 | - | 28 - 29 | 3587 | 119 | |
M55169.1 | - | 27 - 28 | 3376 | 119 | |
M73047.1 | - | 27 - 28 | 3564 | 119 |