The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170715.1 | NP_001164186.1 | 1 - 2 | 139 | 94 |
GenBank mRNA | AK124815.1 | - | 1 - 2 | 142 | 94 |