The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282607.1 | NP_001269536.1 | 10 - 11 | 1488 | 90 |
NM_144967.3 | NP_659404.2 | 10 - 11 | 1724 | 90 | |
XM_006724729.1 | XP_006724792.1 | 9 - 10 | 1195 | 90 | |
XM_011531280.1 | XP_011529582.1 | 9 - 10 | 1212 | 90 | |
GenBank mRNA | AK054620.1 | - | 7 - 8 | 1084 | 90 |
AK128203.1 | - | 10 - 11 | 1724 | 90 | |
AK296748.1 | - | 10 - 11 | 1466 | 90 | |
BC063790.1 | - | 10 - 11 | 1323 | 90 |