The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185059.1 | NP_001171988.1 | 16 - 17 | 3348 | 103 |
NM_024617.3 | NP_078893.2 | 16 - 17 | 3458 | 103 | |
XM_005252207.3 | XP_005252264.1 | 16 - 17 | 3410 | 103 | |
XM_005252208.2 | XP_005252265.1 | 16 - 17 | 3410 | 103 | |
XM_006717283.1 | XP_006717346.1 | 16 - 17 | 3410 | 103 | |
XM_011519011.2 | XP_011517313.1 | 16 - 17 | 3410 | 103 | |
XM_011519012.2 | XP_011517314.1 | 16 - 17 | 3358 | 103 | |
XM_011519013.2 | XP_011517315.1 | 16 - 17 | 4027 | 103 | |
XM_011519014.2 | XP_011517316.1 | 16 - 17 | 3629 | 103 | |
XM_011519015.1 | XP_011517317.1 | 16 - 17 | 3410 | 103 | |
XM_011519016.1 | XP_011517318.1 | 16 - 17 | 3410 | 103 | |
XM_017015130.1 | XP_016870619.1 | 16 - 17 | 4026 | 103 | |
XM_017015131.1 | XP_016870620.1 | 16 - 17 | 3668 | 103 | |
XM_017015132.1 | XP_016870621.1 | 17 - 18 | 3573 | 103 | |
GenBank mRNA | AB051498.1 | - | 8 - 9 | 1996 | 103 |
AK023471.1 | - | 2 - 3 | 103 | ||
AK055948.1 | - | 4 - 5 | 1308 | 103 | |
AL832193.1 | - | 17 - 18 | 3500 | 103 | |
AL832988.1 | - | 3 - 4 | 296 | 103 | |
BC144523.1 | - | 15 - 16 | 3246 | 103 | |
BX641077.1 | - | 15 - 16 | 4808 | 103 | |
CR933643.1 | - | 16 - 17 | 3322 | 103 | |
CR933644.1 | - | 16 - 17 | 3900 | 103 |