The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267576.1 | NP_001254505.1 | 10 - 11 | 1634 | 81 |
NM_153210.4 | NP_694942.3 | 10 - 11 | 1634 | 81 | |
XM_011523639.2 | XP_011521941.1 | 9 - 10 | 1522 | 81 | |
XM_011523640.1 | XP_011521942.1 | 9 - 10 | 1354 | 81 | |
XM_011523641.1 | XP_011521943.1 | 10 - 11 | 1079 | 81 | |
XM_011523642.2 | XP_011521944.1 | 10 - 11 | 1679 | 81 | |
XM_017024159.1 | XP_016879648.1 | 9 - 10 | 1575 | 81 | |
XM_017024160.1 | XP_016879649.1 | 8 - 9 | 1561 | 81 | |
XM_017024161.1 | XP_016879650.1 | 7 - 8 | 1647 | 81 | |
XM_017024162.1 | XP_016879651.1 | 10 - 11 | 1679 | 81 | |
GenBank mRNA | AK055188.1 | - | 10 - 11 | 1115 | 81 |
BC136368.1 | - | 10 - 11 | 1634 | 81 | |
BC144041.1 | - | 10 - 11 | 1634 | 81 | |
BC171759.1 | - | 10 - 11 | 1634 | 81 | |
BX648526.1 | - | 8 - 9 | 897 | 81 |