The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300867.1 | NP_001287796.1 | 10 - 11 | 1130 | 114 |
NM_001324062.1 | NP_001310991.1 | 15 - 16 | 4332 | 114 | |
NM_001324063.1 | NP_001310992.1 | 13 - 14 | 4155 | 114 | |
NM_001324064.1 | NP_001310993.1 | 16 - 17 | 4377 | 114 | |
NM_001324065.1 | NP_001310994.1 | 10 - 11 | 1130 | 114 | |
NM_198551.3 | NP_940953.2 | 15 - 16 | 4332 | 114 | |
XM_006711304.3 | XP_006711367.2 | 13 - 14 | 4467 | 114 | |
XM_011509513.2 | XP_011507815.2 | 15 - 16 | 4644 | 114 | |
XM_017001243.1 | XP_016856732.1 | 15 - 16 | 4157 | 114 | |
GenBank mRNA | AK025122.1 | - | 7 - 8 | 631 | 114 |
AK025506.1 | - | 12 - 13 | 2012 | 114 | |
AK026489.1 | - | 13 - 14 | 1338 | 114 | |
AK096526.1 | - | 10 - 11 | 2893 | 114 | |
AK290335.1 | - | 6 - 7 | 607 | 114 | |
AK296712.1 | - | 10 - 11 | 1111 | 114 | |
AX775845.1 | - | 10 - 11 | 1104 | 114 | |
D87742.1 | - | 12 - 13 | 2181 | 114 |