The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001483.2 | NP_001001483.1 | 10 - 11 | 1197 | 77 |
NM_019028.2 | NP_061901.2 | 11 - 12 | 1343 | 77 | |
XM_005252996.2 | XP_005253053.1 | 11 - 12 | 1315 | 77 | |
XM_011520193.1 | XP_011518495.1 | 9 - 10 | 1040 | 77 | |
XM_011520194.2 | XP_011518496.1 | 11 - 12 | 1318 | 77 | |
XM_011520195.1 | XP_011518497.1 | 11 - 12 | 1253 | 77 | |
GenBank mRNA | AB024495.1 | - | 11 - 12 | 1291 | 77 |
AB097014.1 | - | 11 - 12 | 1341 | 77 | |
AB097027.1 | - | 11 - 12 | 1341 | 77 | |
AK001714.1 | - | 11 - 12 | 1341 | 77 | |
AK001831.1 | - | 6 - 7 | 715 | 77 | |
AK291348.1 | - | 11 - 12 | 1318 | 77 | |
AL833714.1 | - | 7 - 8 | 793 | 77 | |
BC036020.1 | - | 11 - 12 | 1328 | 77 | |
BC050690.1 | - | 10 - 11 | 1197 | 77 | |
BC056152.1 | - | 11 - 12 | 1343 | 77 | |
BX648156.1 | - | 10 - 11 | 1187 | 77 |