The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001483.2 | NP_001001483.1 | 7 - 8 | 829 | 128 |
NM_019028.2 | NP_061901.2 | 8 - 9 | 975 | 128 | |
XM_005252996.2 | XP_005253053.1 | 8 - 9 | 947 | 128 | |
XM_011520193.1 | XP_011518495.1 | 6 - 7 | 672 | 128 | |
XM_011520194.2 | XP_011518496.1 | 8 - 9 | 950 | 128 | |
XM_011520195.1 | XP_011518497.1 | 8 - 9 | 885 | 128 | |
GenBank mRNA | AB024495.1 | - | 8 - 9 | 923 | 128 |
AB097014.1 | - | 8 - 9 | 973 | 128 | |
AB097027.1 | - | 8 - 9 | 973 | 128 | |
AK001714.1 | - | 8 - 9 | 973 | 128 | |
AK001831.1 | - | 3 - 4 | 347 | 128 | |
AK291348.1 | - | 8 - 9 | 950 | 128 | |
AL833714.1 | - | 4 - 5 | 425 | 128 | |
BC036020.1 | - | 8 - 9 | 960 | 128 | |
BC050690.1 | - | 7 - 8 | 829 | 128 | |
BC056152.1 | - | 8 - 9 | 975 | 128 |