The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017919.1 | NP_001017919.1 | 3 - 4 | 758 | 100 |
NM_033544.2 | NP_291022.2 | 4 - 5 | 842 | 100 | |
GenBank mRNA | BC064606.1 | - | 1 - 2 | 769 | 100 |
BC140708.1 | - | 3 - 4 | 756 | 100 | |
BC140709.1 | - | 3 - 4 | 756 | 100 | |
BU850147.1 | - | 2 - 3 | 558 | 100 |