The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014788.3 | NP_055603.2 | 2 - 3 | 461 | 84 |
NM_033219.2 | NP_150088.1 | 2 - 3 | 461 | 84 | |
XM_005252320.3 | XP_005252377.1 | 2 - 3 | 526 | 84 | |
XM_006717338.1 | XP_006717401.1 | 2 - 3 | 1084 | 84 | |
XM_011519244.2 | XP_011517546.1 | 2 - 3 | 291 | 84 | |
XM_017015352.1 | XP_016870841.1 | 2 - 3 | 526 | 84 | |
XM_017015353.1 | XP_016870842.1 | 2 - 3 | 526 | 84 | |
XM_017015354.1 | XP_016870843.1 | 2 - 3 | 517 | 84 | |
GenBank mRNA | AF220130.1 | - | 2 - 3 | 306 | 84 |
AF220131.1 | - | 2 - 3 | 306 | 84 | |
AK055833.1 | - | 2 - 3 | 334 | 84 | |
AK292825.1 | - | 2 - 3 | 328 | 84 | |
BC006333.2 | - | 2 - 3 | 314 | 84 | |
BF797334.1 | - | 2 - 3 | 419 | 84 | |
D50919.1 | - | 2 - 3 | 306 | 84 | |
DC405577.1 | - | 2 - 3 | 461 | 84 |