The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011512262.2 | XP_011510564.1 | 13 - 14 | 1444 | 87 |
XM_011512265.2 | XP_011510567.1 | 11 - 12 | 923 | 87 | |
XM_017005474.1 | XP_016860963.1 | 13 - 14 | 1426 | 87 | |
XM_017005475.1 | XP_016860964.1 | 13 - 14 | 1426 | 87 | |
XM_017005476.1 | XP_016860965.1 | 14 - 15 | 1474 | 87 | |
XM_017005477.1 | XP_016860966.1 | 14 - 15 | 3069 | 87 | |
XM_017005478.1 | XP_016860967.1 | 14 - 15 | 1200 | 87 | |
GenBank mRNA | AK092780.1 | - | 6 - 7 | 1714 | 87 |
BC041437.1 | - | 14 - 15 | 1159 | 87 |