Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT5DC4
Entrez Gene ID
284958
Gene Name
5'-nucleotidase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.2: 112717245 - 112742879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011512262.2 XP_011510564.1 6 - 7 860 70
XM_011512265.2 XP_011510567.1 4 - 5 339 70
XM_017005474.1 XP_016860963.1 6 - 7 860 70
XM_017005475.1 XP_016860964.1 6 - 7 860 70
XM_017005476.1 XP_016860965.1 7 - 8 890 70
XM_017005477.1 XP_016860966.1 7 - 8 2485 70
XM_017005478.1 XP_016860967.1 7 - 8 616 70
GenBank mRNA AK092780.1 - 1 - 2 209 70
BC041437.1 - 7 - 8 593 70

View Full Product Details