Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLD3
Entrez Gene ID
23646
Gene Name
phospholipase D family member 3
Gene Aliases
AD19, HU-K4, HUK4
Chromosome Location
Chr.19: 40348395 - 40378490 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031696.3 NP_001026866.1 4 - 5 506 79
NM_001291311.1 NP_001278240.1 4 - 5 440 79
NM_012268.3 NP_036400.2 4 - 5 730 79
XM_005258704.1 XP_005258761.1 4 - 5 651 79
XM_005258707.4 XP_005258764.1 4 - 5 433 79
XM_005258708.3 XP_005258765.1 3 - 4 326 79
XM_005258709.4 XP_005258766.1 3 - 4 314 79
XM_005258710.4 XP_005258767.1 3 - 4 355 79
XM_006723122.1 XP_006723185.1 4 - 5 818 79
XM_011526692.1 XP_011524994.1 4 - 5 813 79
XM_011526693.1 XP_011524995.1 4 - 5 533 79
XM_017026546.1 XP_016882035.1 5 - 6 749 79
XM_017026547.1 XP_016882036.1 4 - 5 317 79
XM_017026548.1 XP_016882037.1 4 - 5 468 79
XM_017026549.1 XP_016882038.1 4 - 5 571 79
GenBank mRNA AK293690.1 - 3 - 4 231 79
BC036327.1 - 4 - 5 231 79
BC096820.1 - 4 - 5 429 79
BP304863.1 - 3 - 4 246 79
U60644.1 - 4 - 5 437 79

View Full Product Details