The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193286.1 | NP_001180215.1 | 9 - 10 | 981 | 67 |
NM_012237.3 | NP_036369.2 | 10 - 11 | 989 | 67 | |
NM_030593.2 | NP_085096.1 | 9 - 10 | 981 | 67 | |
NR_034146.1 | 8 - 9 | 912 | 67 | ||
XM_006723111.1 | XP_006723174.1 | 11 - 12 | 937 | 67 | |
XM_011526654.1 | XP_011524956.1 | 9 - 10 | 874 | 67 | |
XM_011526655.1 | XP_011524957.1 | 7 - 8 | 689 | 67 | |
XM_017026500.1 | XP_016881989.1 | 9 - 10 | 913 | 67 | |
GenBank mRNA | AB009274.1 | - | 9 - 10 | 702 | 67 |
AF083107.2 | - | 10 - 11 | 887 | 67 | |
AF095714.1 | - | 9 - 10 | 760 | 67 | |
AF131800.1 | - | 8 - 9 | 636 | 67 | |
AF160214.1 | - | 9 - 10 | 741 | 67 | |
AK025876.1 | - | 9 - 10 | 767 | 67 | |
AK054642.1 | - | 10 - 11 | 820 | 67 | |
AK225824.1 | - | 9 - 10 | 764 | 67 | |
AK290716.1 | - | 8 - 9 | 912 | 67 | |
AK314492.1 | - | 10 - 11 | 743 | 67 | |
AY030277.1 | - | 9 - 10 | 742 | 67 | |
BC003012.1 | - | 9 - 10 | 832 | 67 | |
BC003547.1 | - | 9 - 10 | 832 | 67 | |
KF032391.1 | - | 7 - 8 | 689 | 67 |